siRNA Details
| Gene Symbol: Dpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159543.1 | NP_001153015.1 | 20 | 2226 | ||
| NM_010074.3 | NP_034204.1 | 20 | 2226 | |||
| GenBank mRNA | AK085370.1 | - | 20 | 1809 | ||
| AK153939.1 | - | 20 | 1840 | |||
| AK163085.1 | - | 20 | 2231 | |||
| AK164775.1 | - | 20 | 1809 | |||
| BC022183.1 | - | 21 | 2088 | 3982742 | ||
| X58384.1 | - | 20 | 1903 | |||

Dpp4
Genomic Map