siRNA Details
Gene Symbol: Dpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136060.2 | NP_001129532.1 | 6 | 827 | ||
NM_001198886.1 | NP_001185815.1 | 6 | 911 | |||
NM_010075.2 | NP_034205.1 | 6 | 1162 | |||
NM_207282.3 | NP_997165.2 | 6 | 1015 | |||
GenBank mRNA | AF092507.1 | - | 6 | 1170 | ||
AK031903.1 | - | 6 | 788 | |||
AK033808.1 | - | 2 | 119 | |||
AK051708.1 | - | 6 | 993 | |||
AK158964.1 | - | 6 | 1016 | |||
BC048383.1 | - | 6 | 678 | 6306267 | ||
BC085154.1 | - | 6 | 911 | 6838230 |
