siRNA Details
| Gene Symbol: Dpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136060.2 | NP_001129532.1 | 20 | 2093 | ||
| NM_001198886.1 | NP_001185815.1 | 20 | 2177 | |||
| NM_010075.2 | NP_034205.1 | 20 | 2428 | |||
| NM_207282.3 | NP_997165.2 | 20 | 2281 | |||
| GenBank mRNA | AF092506.1 | - | 17 | 1441 | ||
| AF092507.1 | - | 20 | 2436 | |||
| AK031903.1 | - | 20 | 2054 | |||
| AK158964.1 | - | 20 | 2282 | |||
| BC048383.1 | - | 20 | 1944 | 6306267 | ||
| BC085154.1 | - | 20 | 2177 | 6838230 | ||

Dpp6
Genomic Map