siRNA Details
| Gene Symbol: Dr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026106.4 | NP_080382.2 | 1 | 854 | ||
| GenBank mRNA | AK007230.1 | - | 1 | 829 | ||
| AK031057.1 | - | 1 | 788 | |||
| AK075713.1 | - | 1 | 854 | |||
| AK090113.1 | - | 1 | 836 | |||
| AK139866.1 | - | 1 | 784 | |||
| AK144749.1 | - | 1 | 848 | |||
| AK146573.1 | - | 1 | 817 | |||
| AK178588.1 | - | 1 | 74 | |||
| AK211344.1 | - | 1 | 74 | |||
| BC013461.1 | - | 1 | 423 | 4222118 | ||

Dr1
Genomic Map