siRNA Details
| Gene Symbol: Slc26a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021353.3 | NP_067328.1 | 20, 21 | 2472 | ||
| GenBank mRNA | AF136751.1 | - | 20, 21 | 2390 | ||
| AK018617.1 | - | 1 | 898 | |||
| BC139271.1 | - | 19, 20 | 2307 | 8862293 | ||
| BC139273.1 | - | 19, 20 | 2307 | 8862295 | ||

Slc26a3
Genomic Map