siRNA Details
| Gene Symbol: Drd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007877.1 | NP_031903.1 | 6 | 847 | ||
| GenBank mRNA | BC119286.1 | - | 6 | 868 | 8734035 | |
| BC137819.1 | - | 6 | 868 | 8860838 | ||
| X67274.1 | - | 6 | 847 | |||
More Information
Set Membership: |
|

Drd3
Genomic Map