siRNA Details
| Gene Symbol: Drg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007879.1 | NP_031905.1 | 4 | 469 | ||
| GenBank mRNA | AK044194.1 | - | 4 | 478 | ||
| AK080814.1 | - | 4 | 467 | |||
| AK090156.1 | - | 4 | 499 | |||
| AK144438.1 | - | 4 | 471 | |||
| AK155061.1 | - | 4 | 484 | |||
| AK208688.1 | - | 3 | 345 | |||
| BC039649.1 | - | 4 | 463 | 5351885 | ||
| D10715.1 | - | 4 | 469 | |||

Drg1
Genomic Map