siRNA Details
| Gene Symbol: Drp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010078.3 | NP_034208.2 | 19, 20 | 2489 | ||
| GenBank mRNA | AK081426.1 | - | 19, 20 | 2433 | ||
| AK158102.1 | - | 18, 19 | 2291 | |||
| BC060605.1 | - | 17, 18 | 3068 | |||
| BC125345.1 | - | 19, 20 | 2349 | 40129860 | ||
| BC125347.1 | - | 19, 20 | 2349 | 40129862 | ||
| U43520.1 | - | 9, 10 | 1095 | |||

Drp2
Genomic Map