siRNA Details
| Gene Symbol: Dsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291804.1 | NP_001278733.1 | 9 | 1389 | ||
| NM_013504.5 | NP_038532.3 | 9 | 1389 | |||
| GenBank mRNA | AK036893.1 | - | 9 | 1275 | ||
| AK079441.1 | - | 9 | 1391 | |||
| AK081826.1 | - | 9 | 1297 | |||
| BC109161.1 | - | 9 | 1226 | 40047883 | ||
| BC109162.1 | - | 9 | 1226 | |||
| X97986.1 | - | 9 | 1323 | |||

Dsc1
Genomic Map