siRNA Details
| Gene Symbol: Dsc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013505.3 | NP_038533.1 | 6 | 969 | ||
| GenBank mRNA | AK028604.1 | - | 7 | 1009 | ||
| AK028636.1 | - | 6 | 970 | |||
| AK030333.1 | - | 6 | 970 | |||
| AK048547.1 | - | 6 | 970 | |||
| AK172168.1 | - | 6 | 804 | |||
| BC057867.1 | - | 6 | 749 | 5327748 | ||
| L33779.1 | - | 6 | 768 | |||

Dsc2
Genomic Map