siRNA Details
| Gene Symbol: Dscam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031174.4 | NP_112451.1 | 2 | 728 | ||
| GenBank mRNA | AF315558.1 | - | 2 | 373 | ||
| AY005483.1 | - | 2 | 728 | |||
More Information
Other Species Targeted
Set Membership: |
|

Dscam
Genomic Map