siRNA Details
| Gene Symbol: Adam18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010084.2 | NP_034214.2 | 12 | 1191 | ||
| GenBank mRNA | AF167405.1 | - | 12 | 1189 | ||
| AK076633.1 | - | 12 | 1195 | |||
| BC051136.1 | - | 12 | 1168 | 6708158 | ||
| BC144841.1 | - | 12 | 1164 | |||
| BC145751.1 | - | 12 | 1164 | 40131054 | ||

Adam18
Genomic Map