siRNA Details
| Gene Symbol: Adam18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010084.2 | NP_034214.2 | 11 | 989 | ||
| GenBank mRNA | AF167405.1 | - | 11 | 987 | ||
| AK076633.1 | - | 11 | 993 | |||
| BC051136.1 | - | 11 | 966 | 6708158 | ||
| BC144841.1 | - | 11 | 962 | |||
| BC145751.1 | - | 11 | 962 | 40131054 | ||

Adam18
Genomic Map