siRNA Details
| Gene Symbol: Adam24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010086.4 | NP_034216.3 | 2 | 2059 | ||
| GenBank mRNA | AF167402.1 | - | 1 | 2047 | ||
| AK029528.1 | - | 2 | 2060 | |||
| AK133199.1 | - | 2 | 2075 | |||
| BC145785.1 | - | 1 | 1948 | 40131088 | ||

Adam24
Genomic Map