siRNA Details
| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162465.1 | NP_001155937.1 | 11 | 1387 | ||
| NM_007886.2 | NP_031912.2 | 11 | 1322 | |||
| GenBank mRNA | AJ003007.1 | - | 11 | 1255 | ||
| AK043436.1 | - | 11 | 1345 | |||
| AK188811.1 | - | 4 | 371 | |||
| BC030924.1 | - | 11 | 1322 | 4910166 | ||
| Y15742.1 | - | 11 | 1219 | |||

Dtnb
Genomic Map