siRNA Details
Gene Symbol: Dvl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155349.1 | - | 1 | 407 | ||
AK220168.1 | - | 1 | 371 | |||
BC051991.1 | - | 1 | 166 | |||
BC088987.1 | - | 1 | 271 | |||
BC138848.1 | - | 2 | 278 | 8861870 | ||
BC138849.1 | - | 2 | 278 | 8861871 | ||
BU702464.1 | - | 1 | 68 | |||
BY727676.1 | - | 1 | 435 | |||
U10115.1 | - | 1 | 364 |
