siRNA Details
| Gene Symbol: Dvl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155349.1 | - | 15 | 2375 | ||
| AK162430.1 | - | 9 | 1474 | |||
| AK220168.1 | - | 12 | 3820 | |||
| BC018593.1 | - | 8 | 1259 | |||
| BC051991.1 | - | 14 | 2329 | |||
| BC088987.1 | - | 15 | 2349 | |||
| BC138848.1 | - | 16 | 2246 | 8861870 | ||
| BC138849.1 | - | 16 | 2246 | 8861871 | ||
| U10115.1 | - | 15 | 2332 | |||

Dvl1
Genomic Map