siRNA Details
| Gene Symbol: E2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291105.1 | NP_001278034.1 | 7 | 1354 | ||
| NM_007891.5 | NP_031917.1 | 7 | 1398 | |||
| GenBank mRNA | AF483514.1 | - | 7 | 1240 | ||
| AF483515.1 | - | 7 | 1240 | |||
| AK017841.1 | - | 7 | 1346 | |||
| AK153968.1 | - | 7 | 1363 | |||
| AK220226.1 | - | 7 | 1095 | |||
| BC052160.1 | - | 7 | 1340 | 6330425 | ||
| L21973.1 | - | 7 | 1358 | |||

E2f1
Genomic Map