siRNA Details
Gene Symbol: S1pr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010102.2 | NP_034232.1 | 1 | 1147 | ||
GenBank mRNA | AJ006074.1 | - | 1 | 1147 | ||
AK075854.1 | - | 1 | 666 | |||
AK154242.1 | - | 1 | 670 | |||
BC107000.1 | - | 1 | 717 | 40045304 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
