siRNA Details
| Gene Symbol: Edn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007903.4 | NP_031929.1 | 3 | 680 | ||
| GenBank mRNA | AK046164.1 | - | 2 | 642 | ||
| AK048006.1 | - | 3 | 603 | |||
| AK049775.1 | - | 3 | 666 | |||
| AK079150.1 | - | 3 | 679 | |||
| AK162475.1 | - | 2 | 881 | |||
| BC137727.1 | - | 3 | 616 | 8860746 | ||
| BC137730.1 | - | 3 | 616 | 8860749 | ||
| U32330.1 | - | 3 | 820 | |||

Edn3
Genomic Map