siRNA Details
| Gene Symbol: Edn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007903.4 | NP_031929.1 | 2 | 452 | ||
| GenBank mRNA | AK048006.1 | - | 2 | 375 | ||
| AK049775.1 | - | 2 | 438 | |||
| AK079150.1 | - | 2 | 451 | |||
| AK162475.1 | - | 1 | 653 | |||
| BC137727.1 | - | 2 | 388 | 8860746 | ||
| BC137730.1 | - | 2 | 388 | 8860749 | ||
| U32330.1 | - | 2 | 592 | |||

Edn3
Genomic Map