siRNA Details
| Gene Symbol: Eed | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021876.3 | NP_068676.1 | 8 | 1219 | ||
| GenBank mRNA | AK077664.1 | - | 8 | 804 | ||
| AK087924.1 | - | 8 | 780 | |||
| AK131976.1 | - | 8 | 786 | |||
| AK133890.1 | - | 8 | 780 | |||
| BC012966.1 | - | 8 | 1218 | 3991086 | ||
| U78103.1 | - | 8 | 1221 | |||
| U97675.1 | - | 8 | 1226 | |||

Eed
Genomic Map