siRNA Details
| Gene Symbol: Efnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010111.5 | NP_034241.2 | 2 | 428 | ||
| GenBank mRNA | AK132512.1 | - | 2 | 895 | ||
| AK132845.1 | - | 2 | 721 | |||
| AK134108.1 | - | 2 | 895 | |||
| AK142203.1 | - | 2 | 898 | |||
| AK142283.1 | - | 2 | 898 | |||
| AK143059.1 | - | 2 | 895 | |||
| BC057009.1 | - | 2 | 716 | 6827408 | ||
| CT010381.1 | - | 2 | 269 | |||
| L38847.1 | - | 2 | 429 | |||
| U16819.1 | - | 2 | 297 | |||
| U30244.1 | - | 2 | 266 | |||

Efnb2
Genomic Map