siRNA Details
| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007912.4 | NP_031938.1 | 4 | 751 | ||
| NM_207655.2 | NP_997538.1 | 4 | 751 | |||
| GenBank mRNA | AF124513.1 | - | 4 | 685 | ||
| AF275367.1 | - | 4 | 694 | |||
| AK004883.1 | - | 4 | 752 | |||
| AK004911.1 | - | 4 | 726 | |||
| AK004944.1 | - | 4 | 745 | |||
| AK134262.1 | - | 4 | 725 | |||
| BC023729.1 | - | 4 | 722 | 5351807 | ||
| U03425.1 | - | 4 | 521 | |||
| X59698.1 | - | 4 | 630 | |||
| X78987.1 | - | 4 | 530 | |||

Egfr
Genomic Map