siRNA Details
Gene Symbol: Ei24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199494.1 | NP_001186423.1 | 6 | 609 | ||
NM_007915.5 | NP_031941.1 | 6 | 493 | |||
GenBank mRNA | AK145018.1 | - | 6 | 570 | ||
AK147776.1 | - | 6 | 522 | |||
AK159927.1 | - | 6 | 551 | |||
AK172233.1 | - | 6 | 573 | |||
AK212828.1 | - | 1 | 54 | |||
BU148129.1 | - | 6 | 562 | |||
U41751.2 | - | 6 | 497 |
