siRNA Details
| Gene Symbol: Eif2b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127355.1 | NP_001120827.1 | 10 | 1220 | ||
| NM_001127356.1 | NP_001120828.1 | 9 | 1442 | |||
| NM_010122.2 | NP_034252.2 | 10 | 1075 | |||
| GenBank mRNA | AK154364.1 | - | 10 | 983 | ||
| AK158282.1 | - | 10 | 984 | |||
| BC021884.1 | - | 10 | 962 | 4168186 | ||
| M98035.1 | - | 9 | 1062 | |||
| M98036.1 | - | 10 | 1028 | |||

Eif2b4
Genomic Map