siRNA Details
| Gene Symbol: Elf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286411.1 | NP_001273340.1 | 9 | 1961 | ||
| NM_001286412.1 | NP_001273341.1 | 8 | 1850 | |||
| NM_007920.4 | NP_031946.1 | 9 | 2308 | |||
| GenBank mRNA | AK132456.1 | - | 9 | 1962 | ||
| AK142807.1 | - | 6 | 1367 | |||
| AK156098.1 | - | 8 | 1840 | |||
| BC057134.1 | - | 9 | 2308 | 6853870 | ||
| U19617.1 | - | 9 | 1955 | |||

Elf1
Genomic Map