siRNA Details
| Gene Symbol: Elk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013508.2 | NP_038536.2 | 3 | 1245 | ||
| GenBank mRNA | AK052546.1 | - | 3 | 922 | ||
| AK133847.1 | - | 3 | 941 | |||
| BC005686.1 | - | 3 | 1227 | 3155850 | ||
| BC053402.1 | - | 3 | 996 | 30060743 | ||
| BC055735.1 | - | 3 | 1018 | 6808738 | ||
| L19953.1 | - | 3 | 1136 | |||
| S82864.1 | - | 3 | 928 | |||
| Z32815.1 | - | 3 | 1129 | |||

Elk3
Genomic Map