siRNA Details
| Gene Symbol: Enah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083120.2 | NP_001076589.1 | 9, 10 | 2515 | ||
| NM_001083121.2 | NP_001076590.1 | 9, 10 | 1783 | |||
| NM_008680.3 | NP_032706.2 | 10, 11 | 2527 | |||
| NM_010135.3 | NP_034265.2 | 10, 11 | 2572 | |||
| GenBank mRNA | BC062927.1 | - | 9, 10 | 1402 | 6851217 | |
| U72520.1 | - | 9, 10 | 1476 | |||
| U72521.1 | - | 9, 10 | 2202 | |||
| U72522.1 | - | 10, 11 | 2214 | |||
| U72523.1 | - | 10, 11 | 2259 | |||

Enah
Genomic Map