siRNA Details
| Gene Symbol: Enam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017468.3 | NP_059496.1 | 10 | 3427 | ||
| GenBank mRNA | BC115848.1 | - | 10 | 3339 | 40048943 | |
| U82698.1 | - | 10 | 3428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Enam| Gene Symbol: Enam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017468.3 | NP_059496.1 | 10 | 3427 | ||
| GenBank mRNA | BC115848.1 | - | 10 | 3339 | 40048943 | |
| U82698.1 | - | 10 | 3428 | |||