siRNA Details
Gene Symbol: Enam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017468.3 | NP_059496.1 | 7 | 735 | ||
GenBank mRNA | BC115848.1 | - | 7 | 647 | 40048943 | |
U82698.1 | - | 7 | 736 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Enam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017468.3 | NP_059496.1 | 7 | 735 | ||
GenBank mRNA | BC115848.1 | - | 7 | 647 | 40048943 | |
U82698.1 | - | 7 | 736 |