siRNA Details
| Gene Symbol: Eng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146348.1 | NP_001139820.1 | 14 | 2174 | ||
| NM_001146350.1 | NP_001139822.1 | 14 | 2177 | |||
| NM_007932.2 | NP_031958.2 | 14 | 2177 | |||
| GenBank mRNA | AK018588.1 | - | 2 | 137 | ||
| AK151302.1 | - | 14 | 2147 | |||
| AK178966.1 | - | 3 | 186 | |||
| AY679531.1 | - | 14 | 2177 | |||
| BC029080.1 | - | 14 | 2091 | 4168250 | ||
| S69407.1 | - | 15 | 2042 | |||
| X77952.1 | - | 14 | 2177 | |||

Eng
Genomic Map