siRNA Details
| Gene Symbol: Enpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007934.3 | NP_031960.1 | 1 | 720 | ||
| GenBank mRNA | AK034613.1 | - | 1 | 692 | ||
| AK128975.1 | - | 1 | 677 | |||
| AK146808.1 | - | 1 | 692 | |||
| BC127060.1 | - | 1 | 584 | 40109863 | ||
| DQ000292.1 | - | 1 | 460 | |||
| M29961.1 | - | 1 | 539 | |||

Enpep
Genomic Map