siRNA Details
| Gene Symbol: Epb4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013513.2 | NP_038541.1 | 9 | 1318 | ||
| GenBank mRNA | AK133896.1 | - | 9 | 1332 | ||
| AK137484.1 | - | 9 | 1399 | |||
| AK143841.1 | - | 9 | 1298 | |||
| AK165288.1 | - | 9 | 1245 | |||
| BC066194.1 | - | 9 | 1318 | 30429971 | ||
| BC066806.1 | - | 9 | 1318 | 30429961 | ||
| L35933.1 | - | 9 | 1297 | |||
| U03487.1 | - | 9 | 1248 | |||
| U04055.1 | - | 9 | 1318 | |||

Epb4.2
Genomic Map