siRNA Details
| Gene Symbol: Stom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013515.2 | NP_038543.1 | 7 | 797 | ||
| GenBank mRNA | AF093620.1 | - | 7 | 748 | ||
| AK139242.1 | - | 7 | 765 | |||
| AK143724.1 | - | 7 | 765 | |||
| AK148975.1 | - | 7 | 766 | |||
| AK149689.1 | - | 7 | 765 | |||
| AK149821.1 | - | 7 | 765 | |||
| AK149991.1 | - | 7 | 765 | |||
| AK151129.1 | - | 7 | 764 | |||
| U17297.1 | - | 7 | 797 | |||

Stom
Genomic Map