siRNA Details
| Gene Symbol: Epc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007935.2 | NP_031961.1 | 6 | 968 | ||
| NM_027497.3 | NP_081773.1 | 6 | 1119 | |||
| GenBank mRNA | AF079765.1 | - | 6 | 947 | ||
| AK040254.1 | - | 6 | 1119 | |||
| AK148194.1 | - | 6 | 937 | |||
| AK149317.1 | - | 3 | 319 | |||
| BC138622.1 | - | 6 | 947 | 8861642 | ||
| BC138623.1 | - | 6 | 947 | 8861643 | ||

Epc1
Genomic Map