siRNA Details
| Gene Symbol: Epha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023580.4 | NP_076069.2 | 12 | 2065 | ||
| GenBank mRNA | AF131197.1 | - | 12 | 2007 | ||
| AK028478.1 | - | 12 | 2066 | |||
| BC071215.1 | - | 12 | 2029 | 4196138 | ||
| U18084.1 | - | 8 | 1168 | |||

Epha1
Genomic Map