siRNA Details
| Gene Symbol: Epha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007936.3 | NP_031962.2 | 5 | 1310 | ||
| GenBank mRNA | AK028509.1 | - | 5 | 1303 | ||
| AK132203.1 | - | 5 | 1309 | |||
| AK147698.1 | - | 5 | 1306 | |||
| BC052164.1 | - | 5 | 1282 | 6512978 | ||
| S57168.1 | - | 5 | 1310 | |||
| X65138.1 | - | 5 | 1310 | |||

Epha4
Genomic Map