siRNA Details
| Gene Symbol: Epha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122889.1 | NP_001116361.1 | 7 | 1830 | ||
| NM_001290434.1 | NP_001277363.1 | 7 | 1844 | |||
| NM_010141.4 | NP_034271.3 | 7 | 1844 | |||
| GenBank mRNA | AK030480.1 | - | 7 | 1811 | ||
| AK033903.1 | - | 7 | 1845 | |||
| AK049848.1 | - | 7 | 1811 | |||
| BC026153.1 | - | 7 | 1736 | 3991628 | ||
| X79082.1 | - | 7 | 1823 | |||
| X79083.1 | - | 7 | 1823 | |||
| X79084.1 | - | 7 | 1823 | |||
| X81466.1 | - | 3 | 301 | |||

Epha7
Genomic Map