siRNA Details
| Gene Symbol: Ephb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159571.1 | NP_001153043.1 | 10 | 2236 | ||
| NM_010144.6 | NP_034274.4 | 10 | 2209 | |||
| GenBank mRNA | AK046884.1 | - | 10 | 2174 | ||
| AK049340.1 | - | 10 | 2210 | |||
| AK132392.1 | - | 10 | 2209 | |||
| AK143422.1 | - | 2179 | ||||
| BC013342.1 | - | 10 | 1897 | |||
| BC016505.1 | - | 10 | 2161 | 3709926 | ||
| BC090839.1 | - | 10 | 1986 | |||
| U06834.1 | - | 11 | 1822 | |||

Ephb4
Genomic Map