siRNA Details
| Gene Symbol: Epm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010146.2 | NP_034276.2 | 3 | 546 | ||
| GenBank mRNA | AF124044.1 | - | 3 | 586 | ||
| BY730431.1 | - | 3 | 547 | |||
| CN532498.1 | - | 2 | 177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epm2a| Gene Symbol: Epm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010146.2 | NP_034276.2 | 3 | 546 | ||
| GenBank mRNA | AF124044.1 | - | 3 | 586 | ||
| BY730431.1 | - | 3 | 547 | |||
| CN532498.1 | - | 2 | 177 | |||