siRNA Details
| Gene Symbol: Eps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159964.1 | NP_001153436.1 | 12 | 1600 | ||
| NM_007943.3 | NP_031969.1 | 24 | 2549 | |||
| GenBank mRNA | AK083176.1 | - | 12 | 1584 | ||
| BC048783.1 | - | 24 | 2523 | 6466448 | ||
| L21768.1 | - | 24 | 2522 | |||

Eps15
Genomic Map