siRNA Details
| Gene Symbol: Eps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159964.1 | NP_001153436.1 | 2 | 320 | ||
| NM_007943.3 | NP_031969.1 | 14 | 1269 | |||
| GenBank mRNA | AK028361.1 | - | 123 | |||
| AK083176.1 | - | 2 | 304 | |||
| AY190610.1 | - | 14 | 1242 | |||
| BC048783.1 | - | 14 | 1243 | 6466448 | ||
| CF533628.1 | - | 2 | 320 | |||
| L21768.1 | - | 14 | 1242 | |||

Eps15
Genomic Map