siRNA Details
| Gene Symbol: Eps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271587.1 | NP_001258516.1 | 15 | 1859 | ||
| NM_001271588.1 | NP_001258517.1 | 15 | 1790 | |||
| NM_001271589.1 | NP_001258518.1 | 8 | 860 | |||
| NM_001271595.1 | NP_001258524.1 | 15 | 1864 | |||
| NM_007945.3 | NP_031971.2 | 16 | 1865 | |||
| GenBank mRNA | AK136246.1 | - | 6 | 631 | ||
| AK147877.1 | - | 16 | 1793 | |||
| AK149683.1 | - | 15 | 1653 | |||
| AK155022.1 | - | 15 | 1625 | |||
| AK165684.1 | - | 8 | 863 | |||
| AK166156.1 | - | 16 | 1700 | |||
| BC016890.1 | - | 15 | 1876 | 4240899 | ||
| L21671.1 | - | 15 | 1780 | |||

Eps8
Genomic Map