siRNA Details
Gene Symbol: Ercc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133658.1 | NP_598419.1 | 10 | 1701 | ||
GenBank mRNA | AK151188.1 | - | 10 | 1706 | ||
AK152347.1 | - | 10 | 1706 | |||
AK152988.1 | - | 10 | 1713 | |||
AK154745.1 | - | 10 | 1707 | |||
AK160187.1 | - | 4 | 805 | |||
BC016595.1 | - | 10 | 1672 | 3153847 | ||
BC022956.1 | - | 9 | 1805 | |||
BC024446.1 | - | 1 | 47 | |||
BC026575.1 | - | 10 | 1683 | 4955015 | ||
S71186.1 | - | 10 | 1701 |
