siRNA Details
| Gene Symbol: Ercc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133658.1 | NP_598419.1 | 10, 11 | 1790 | ||
| GenBank mRNA | AK151188.1 | - | 10, 11 | 1795 | ||
| AK152347.1 | - | 10, 11 | 1795 | |||
| AK152988.1 | - | 10, 11 | 1802 | |||
| AK154745.1 | - | 10, 11 | 1796 | |||
| AK160187.1 | - | 4, 5 | 894 | |||
| BC016595.1 | - | 10, 11 | 1761 | 3153847 | ||
| BC022956.1 | - | 9, 10 | 1894 | |||
| BC024446.1 | - | 1, 2 | 136 | |||
| BC026575.1 | - | 10, 11 | 1772 | 4955015 | ||
| S71186.1 | - | 10, 11 | 1790 | |||

Ercc3
Genomic Map