siRNA Details
| Gene Symbol: Esr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010157.3 | NP_034287.3 | 8 | 1714 | ||
| NM_207707.1 | NP_997590.1 | 9 | 1768 | |||
| GenBank mRNA | AK054290.1 | - | 9 | 1766 | ||
| AK054301.1 | - | 7 | 1576 | |||
| AK054413.1 | - | 9 | 1769 | |||
| AY054413.1 | - | 8 | 1229 | |||
| BC141075.1 | - | 7 | 1416 | 9055687 | ||
| BC145329.1 | - | 7 | 1416 | 9053885 | ||
| U81451.1 | - | 7 | 1186 | |||

Esr2
Genomic Map