siRNA Details
| Gene Symbol: Esr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010157.3 | NP_034287.3 | 2 | 485 | ||
| NM_207707.1 | NP_997590.1 | 2 | 485 | |||
| GenBank mRNA | AF063853.1 | - | 1 | 81 | ||
| AF067422.1 | - | 1 | 138 | |||
| AK054290.1 | - | 2 | 483 | |||
| AK054301.1 | - | 2 | 486 | |||
| AK054413.1 | - | 2 | 486 | |||
| BC141075.1 | - | 1 | 187 | 9055687 | ||
| BC145329.1 | - | 1 | 187 | 9053885 | ||

Esr2
Genomic Map