siRNA Details
| Gene Symbol: Mecom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007963.2 | NP_031989.2 | 8 | 1838 | ||
| GenBank mRNA | AJ001482.1 | - | 6 | 1966 | ||
| AK033517.1 | - | 8 | 1272 | |||
| BC076620.1 | - | 8 | 1790 | |||
| BC139762.1 | - | 6 | 956 | 40109903 | ||
| JQ665270.1 | - | 6 | 1958 | |||

Mecom
Genomic Map