siRNA Details
| Gene Symbol: Mecom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007963.2 | NP_031989.2 | 7 | 1667 | ||
| GenBank mRNA | AJ001482.1 | - | 5 | 1795 | ||
| AK033517.1 | - | 7 | 1101 | |||
| BC076620.1 | - | 7 | 1619 | |||
| BC139762.1 | - | 5 | 785 | 40109903 | ||
| JQ665270.1 | - | 5 | 1787 | |||

Mecom
Genomic Map